Control Slides, Special Stains, Amyloid Or Congo Red
Supplier: Mortech Manufacturing
SC802-10 SC802-100 SC802-20
76947-826CS 416.76 CAD
76947-826 76947-828 76947-830
Control Slides, Special Stains, Amyloid Or Congo Red
Slides Microscopy Control Slides
All positive control slides are on Mopec IHC enhanced hydrophilic adhesive slides using formalin fixed paraffin embedded (FFPE) human tissue.
  • Revalidate after one year to verify continued reactivity
  • Amyloid controls are cut in serial sections at 8 µm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Packaging: Case of 10 contains 10 unstained slides and 1 stained slide. Case of 20 contains 20 unstained slides and 2 stained slides. Case of 100 contains 98 unstained slides and 2 stained slides.