Control Slides IHC Mammaglobin
Supplier: Mortech Manufacturing
SC333-100 SC333-25
76946-668CS 2554.56 CAD
76946-668 76946-670
Control Slides IHC Mammaglobin
Slides Microscopy Control Slides
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using formalin fixed paraffin embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properlyMore Product Information
Order Now
SPECIFICATIONS
Species
Environmentally Preferable