Control Slides, Special Stains, Warthin-Starry Stain, Legionella or H. Pylori
Supplier: Mortech Manufacturing
SC834-10 SC834-100 SC834-20
76948-018CS 416.76 CAD
76948-018 76948-020 76948-022
Control Slides, Special Stains, Warthin-Starry Stain, Legionella or H. Pylori
Slides Microscopy Control Slides
All positive control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 μm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Ordering information: Case of 10 contains 10 unstained slides and 1 stained slide. Case of 20 contains 20 unstained slides and 2 stained slides. Case of 100 contains 98 unstained slides and 2 stained slides.