Control Slides IHC MUM1 IRF4
Supplier: Mortech Manufacturing
SC350-100 SC350-25
76946-736CS 2554.56 CAD
76946-736 76946-738
Control Slides IHC MUM1 IRF4
Slides Microscopy Control Slides
All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 μm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly atMore Product Information
Order Now
SPECIFICATIONS
Species
Environmentally Preferable