Microscope Slides, IHC, SuperWhite Glass, Hydrophilic, 90° Corners
Supplier: Mortech Manufacturing
SL390-BL-BX SL390-BL-CS SL390-PK-BX SL390-PK-CS SL390-WH-BX SL390-WH-CS
76948-450BX 113.85 CAD
76948-450 76948-452 76948-454 76948-456 76948-458 76948-460
Microscope Slides, IHC, SuperWhite Glass, Hydrophilic, 90° Corners
Slides Microscope Slides
Charged SuperWhite glass microscope slides for use in immunohistochemistry.
  • Compatible with most auto-stainers, including Ventana
  • Standard size 75×25 mm, with 1 mm thickness
  • Hydrophilic glass withstands extreme staining environments
  • Strong electrostatically charged surface
  • Pure white, ultra-clear glass
Slides are available in white, blue, and pink. The packaging options include 1 box of 72 slides and 1 case of 1440 slides.