Control Slides, Normal Bone Tissue
Supplier: Mortech Manufacturing
SC952-100 SC952-25
76948-246CS 2025.47 CAD
76948-246 76948-248
Control Slides, Normal Bone Tissue
Slides Microscopy Control Slides
All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 microns
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when storedMore Product Information
Order Now
SPECIFICATIONS
Species
Environmentally Preferable