Control Slides IHC PEGFR
Supplier: Mortech Manufacturing
SC423-100 SC423-25
76947-028CS 2554.56 CAD
76947-028 76947-030
Control Slides IHC PEGFR
Slides Microscopy Control Slides
All control slides are on Mopec IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 microns
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Packaging: Case of 25 contains 24 unstained slides and 1 stained slide. Case of 100 contains 98 unstained slides and 2 stained slides.