Specimen Chucks for CRYOSTAR Cryostats, 40 mm, Available in Multiple Colors
Supplier: Mortech Manufacturing
SP124-BL SP124-GN SP124-OR SP124-PK SP124-PR SP124-RD SP124W-BL SP124W-GN SP124-WH SP124W-OR SP124W-PK SP124W-PR SP124W-RD SP124W-WH SP124W-YE SP124-YE
76948-692EA 363.86 CAD
76948-692 76948-694 76948-696 76948-698 76948-700 76948-702 76948-724 76948-726 76948-704 76948-728 76948-730 76948-732 76948-734 76948-736 76948-738 76948-706
Specimen Chucks for CRYOSTAR Cryostats, 40 mm, Available in Multiple Colors
Specimen Mounting Supplies
Available in multiple sizes and colors. Some variations also available in waffle pattern.
  • Round, aluminum specimen chucks
  • Suitable for CRYOSTAR cryostats
  • 40 mm
Available in red, orange, yellow, green, blue, purple, pink and white.