Control Slides, Special Stains, Copper
Supplier: Mortech Manufacturing
SC809-10 SC809-100 SC809-20
76947-868CS 553.87 CAD
76947-868 76947-870 76947-872
Control Slides, Special Stains, Copper
Slides Microscopy Control Slides
All positive control slides are on Mopec IHC enhanced hydrophilic adhesive slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 microns
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt whenMore Product Information
Order Now
SPECIFICATIONS
Species
Environmentally Preferable