Control Slides IHC KI67/DESMIN
Supplier: Mortech Manufacturing
SC292-100 SC292-25
76946-504CS 2857.34 CAD
76946-504 76946-506
Control Slides IHC KI67/DESMIN
Slides Microscopy Control Slides
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µ
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 6More Product Information
Order Now
SPECIFICATIONS
Species
Environmentally Preferable