Anti-Claudin 1 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP153-01-RUO RP153-05-RUO RP153-RUO
CA76635-156EA 141.47 CAD
CA76635-156 CA76635-158 CA76635-160
Anti-Claudin 1 Rabbit Polyclonal Antibody
Antibodies
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact, made up of a network of strands to act as a molecular gasket for preventing the leakage of ions, water etc., between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different claudin composition.