Anti-Helicobacter pylori Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP169-01-RUO RP169-05-RUO RP169-RUO
CA76635-180EA 127.93 CAD
CA76635-180 CA76635-182 CA76635-184
Anti-Helicobacter pylori Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with the 54 kDa (flagellin) of H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.