Anti-Phosphohistone H3 (PHH3) Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems
RP168-05-RUO RP168-RUO RP168-01-RUO
CA76635-176EA 355.18 CAD
CA76635-176 CA76635-178 CA76635-174
Anti-Phosphohistone H3 (PHH3) Rabbit Polyclonal Antibody
Antibodies
Phosphohistone H3 (PHH3) is a core histone protein, which together with other histones, forms the major protein constituents of the chromatin in eukaryotic cells. In mammalian cells, phosphohistone H3 is negligible during interphase but reaches a maximum for chromatin condensation during mitosis.Immunohistochemical studies showed anti-PHH3 specifically detected the core protein histone H3 only when phosphorylated at serine 10 or serine 28. Studies have also revealed no phosphorylation on the histone H3 during apoptosis.PHH3 can serve as a mitotic marker to separate mitotic figures fromMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol PHH3
Antigen name Phosphohistone H3
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR