Anti-Bob1 Rabbit Monoclonal Antibody [clone: SP92]
Supplier: Diagnostic Biosystems
RMAB110-01-RUO RMAB110-05-RUO RMAB110-RUO
CA76634-832EA 200.19 CAD
CA76634-832 CA76634-834 CA76634-836
Anti-Bob1 Rabbit Monoclonal Antibody [clone: SP92]
Antibodies
Transcriptional coactivator that specifically associates with either OCT1 or OCT2. It boosts the OCT1 mediated promoter activity and to a lesser extent, that of OCT2. It has no intrinsic DNAbinding activity. It recognizes the POU domains of OCT1 and OCT2. It is essential for the response of B-cells to antigens and required for the formation of germinal centers.