Anti-PRAME Rabbit Monoclonal Antibody [clone: EPR20330]
Supplier: Diagnostic Biosystems
RMAB109-01-RUO RMAB109-05-RUO RMAB109-RUO
CA76634-826EA 385.28 CAD
CA76634-826 CA76634-828 CA76634-830
Anti-PRAME Rabbit Monoclonal Antibody [clone: EPR20330]
Antibodies
PRAME (Preferentially expressed Antigen in Melanoma) is a tumorassociated antigen and is a member of the family of cancer testis antigens (CTA). PRAME is expressed in malignant cells, including leukaemias, Hodgkin's lymphoma, breast cancer, and primary and metastatic melanomas. PRAME has low or no expression in normal tissues except for in testis, ovary, placenta, adrenals and endometrium.