Anti-PLAP Rabbit Monoclonal Antibody [clone: SP15]
Supplier: Diagnostic Biosystems
RMAB017-01-RUO RMAB017-05-RUO RMAB017-RUO
CA76634-756EA 186.63 CAD
CA76634-756 CA76634-758 CA76634-760
Anti-PLAP Rabbit Monoclonal Antibody [clone: SP15]
Antibodies
This antibody reacts with a membrane-bound isoenzyme (Regan and Nagao type) of placental alkaline phosphatase (PLAP) occurring in the placenta during the 3rd trimester of gestation. This antibody is highly specific to PLAP and shows no cross-reaction with other isoenzymes of alkaline phosphatases. It is useful in the identification of testicular germ cell tumors. Unlike germ cell tumors, PLAP-positive somatic cell tumors uniformly express epithelial membrane antigen (EMA).