Anti-Mmp-9 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP066-01-RUO RP066-05-RUO RP066-RUO
CA76635-084EA 168.55 CAD
CA76635-084 CA76635-086 CA76635-088
Anti-Mmp-9 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 92 kDa pro form and 86 kDa active form of MMP-9. MMPs are proteolytic enzymes capable of degrading connective tissue components. MMPs play a crucial role in tumor cell invasion and metastasis.