Anti-Cd117/c-kit Mouse Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP063-01-RUO RP063-05-RUO RP063-RUO
CA76635-078EA 127.93 CAD
CA76635-078 CA76635-080 CA76635-082
Anti-Cd117/c-kit Mouse Polyclonal Antibody
Antibodies
This antibody reacts with a 145 kDa protein. CD117/c-kit is a transmembrane receptor tyrosine kinase and is expressed in many tissues and cells. CD117/c-kit is involved in the development of several lineages of stem cells, such as germ cells, neural crest derived melanocytes and hematopoietic precursor cells. This antibody can be used in the identification of gastrointestinal stromal tumors.