Anti-Myeloperoxidase Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP053-01-RUO RP053-05-RUO RP053-RUO
CA76635-066EA 127.93 CAD
CA76635-066 CA76635-068 CA76635-070
Anti-Myeloperoxidase Rabbit Polyclonal Antibody
Antibodies
Myeloperoxidase metabolizes most of the hydrogen peroxide generated by activated phagocytes. This antibody stains strongly with the cytoplasm of myeloid cells. The antibody detects myeloperoxidase found in myeloid cells of acute myeloid leukemia. The myeloperoxidase gene has been localized to chromosome 17.