Anti-Mesothelian Rabbit Monoclonal Antibody [clone: YP158]
Supplier: Diagnostic Biosystems
RMAB107-01-RUO RMAB107-05-RUO RMAB107-RUO
CA76634-820EA 135.45 CAD
CA76634-820 CA76634-822 CA76634-824
Anti-Mesothelian Rabbit Monoclonal Antibody [clone: YP158]
Antibodies
Mesothelin is a glycocylphosphotidylianositol-linked cell-surface glycoprotein, which is present on the surface of normal mesothelium and is overexpressed in many patients with epithelial ovarian cancer and malignant mesotheliomas.