Anti-P63 Rabbit Monoclonal Antibody [clone: DBR16.1]
Supplier: Diagnostic Biosystems
RMAB086-01-RUO RMAB086-05-RUO RMAB086-RUO
CA76634-808EA 158.03 CAD
CA76634-808 CA76634-810 CA76634-812
Anti-P63 Rabbit Monoclonal Antibody [clone: DBR16.1]
Antibodies
p63 is a homolog of the tumor suppressor p53, it is identified in basal cells in the epithelial layers of a variety of tissues, including epidermis, cervix, urothelium, breast and prostate. p63, vital for the development of the prostate and selectively expressed by normal prostate basal cells, has been shown to be a promising complementary basal cell specific marker to High Molecular Weight-Cytokeratin (HMW-CK), for the differential diagnosis of benign prostatic lesions and prostatic carcinoma. p63 has also been shown to be a sensitive marker for lung squamous cell carcinomas (SqCC), with a sensitivity of ~90% . Specificity for lung SqCC, vs. lung adenocarcinoma (LADC), is approximately 80. In breast tissue, p63 has been identified in myoepithelial cells of normal ducts.