Anti-CD23 B Cell Rabbit Monoclonal Antibody [clone: SP23]
Supplier: Diagnostic Biosystems
RMAB013-01-RUO RMAB013-05-RUO RMAB013-RUO
CA76634-744EA 213.71 CAD
CA76634-744 CA76634-746 CA76634-748
Anti-CD23 B Cell Rabbit Monoclonal Antibody [clone: SP23]
Antibodies
This antibody recognizes CD23. CD23 is a 45 kDa glycoprotein, which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukemia and some cases of centroblastic/centrocytic lymphoma.