Anti-Gastrin Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP044-01-RUO RP044-05-RUO RP044-RUO
CA76635-030EA 127.93 CAD
CA76635-030 CA76635-032 CA76635-034
Anti-Gastrin Rabbit Polyclonal Antibody
Antibodies
The antibody is specific against gastrin-containing cells. This anti-body reacts with human gastrin I and human gastrin I fragments. No cross-reactivity was observed with rat gastrin, human big gastrin, human gastrin releasing peptide, sulfated cholecystokinin amide, non-sulfated cholecystokinin amide and cholecystokinin.