Anti-Estrogen Receptor (ER) Rabbit Monoclonal Antibody [clone: SP1]
Supplier: Diagnostic Biosystems
RMAB001-01-RUO RMAB001-05-RUO RMAB001-RUO
CA76634-690EA 430.44 CAD
CA76634-690 CA76634-692 CA76634-694
Anti-Estrogen Receptor (ER) Rabbit Monoclonal Antibody [clone: SP1]
Antibodies
This antibody recognizes a protein of 67 kDa, which is identified as estrogen receptor (ER). ER content of breast cancer tissue is an important parameter in the prediction of prognosis and response to endocrine therapy. This antibody strongly stains the nucleus of epithelial cells in breast carcinomas. The ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.