Anti-Glut-1 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP128-01-RUO RP128-05-RUO RP128-RUO
CA76635-144EA 165.55 CAD
CA76635-144 CA76635-146 CA76635-148
Anti-Glut-1 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 55 kDa protein. Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response in insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus and breast. GLUT-1 immunoreactivity in some cancers, including trans-carcinoma of the urinary bladder, has been associated with aggressive behavior.