Anti-IgE Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP022-01-RUO RP022-05-RUO RP022-RUO
CA76634-970EA 127.93 CAD
CA76634-970 CA76634-972 CA76634-974
Anti-IgE Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with the ε chain of human IgE. Traces of contaminating antibodies have been removed by solid phase adsorption. Only the IgE arch appeared in crossed immunoelectrophoresis.