Anti-P16 Mouse Antibody [clone: JC2]
Supplier: Diagnostic Biosystems
MOB575-01-RUO MOB575-05-RUO MOB575-RUO
CA76634-180EA 190.77 CAD
CA76634-180 CA76634-182 CA76634-184
Anti-P16 Mouse Antibody [clone: JC2]
Antibodies
P16 is a mitotic inhibitor protein. It competes with D-type cyclins to bind to cdk4 and cdk6. It acts as tumor suppressor and inhibits the progression of cells through the G1 phase of the cell cycle.