Anti-PTEN Mouse Monoclonal Antibody [clone: 6H2.1]
Supplier: Diagnostic Biosystems
MOB574-05-RUO MOB574-RUO MOB574-01-RUO
CA76634-176EA 752.85 CAD
CA76634-176 CA76634-178 CA76634-174
Anti-PTEN Mouse Monoclonal Antibody [clone: 6H2.1]
Antibodies
PTEN gene is a tumor suppressor gene that maps to chromosome 10q23. PTEN, a novel tumor suppressor, functions as a regulator of both cell cycle progression and apoptosis ). Potentially, mutation and deletion of PTEN gene may result in a new signal transduction pathway related to human malignant tumors. Studies have demonstrated a reduction of PTEN expression in advanced breast cancers.