Anti-Pancreatic polypeptide Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems
RP030-RUO RP030-01-RUO RP030-05-RUO
CA76635-016EA 976.79 CAD
CA76635-016 CA76635-012 CA76635-014
Anti-Pancreatic polypeptide Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with pancreatic polypeptide. It stains the periphery of islets, exocrine pancreatic parenchyma and the epithelium of small and medium sized ducts and acinar cells.
Order Now

SPECIFICATIONS

Antigen name Pancreatic polypeptide
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR