Anti-Lysozyme Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP028-01-RUO RP028-05-RUO RP028-RUO
CA76635-006EA 127.93 CAD
CA76635-006 CA76635-008 CA76635-010
Anti-Lysozyme Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with lysozyme. It stains granulocytes, monocytes and macrophages in human tonsil, skin and colon. This antibody does not cross-react with any other cell types.