Anti-Fibronectin Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP013-05-RUO RP013-RUO RP013-01-RUO
CA76634-936EA 918.07 CAD
CA76634-936 CA76634-938 CA76634-934
Anti-Fibronectin Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human fibronectin in liver, tonsil, skin and kidney. Traces of contaminating antibodies have been removed by solid-phase adsorption. Only the fibronectin arch was observed in crossed immunoelectrophoresis.