Anti-Factor VIII R antigen Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP012-01-RUO RP012-05-RUO RP012-RUO
CA76634-928EA 186.63 CAD
CA76634-928 CA76634-930 CA76634-932
Anti-Factor VIII R antigen Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human factor VIII related antigen. It stains endothelial cells and megakaryocytes in various human tissues. This antibody does not cross-react with any other cell types.