Anti-Uroplakin II (UPK2) Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
PDR177-RUO
CA76634-682EA 556.87 CAD
CA76634-682
Anti-Uroplakin II (UPK2) Rabbit Polyclonal Antibody
Antibodies
Uroplakins (UPs) are a family of transmembrane proteins (UPs Ia, Ib, II and III) that are specific differentiation products of urothelial cells. Uroplakins are markers of terminally differentiated urothelium. Uroplakin II (UPII) is a newly described sensitive marker for urothelial carcinoma (UC). The expression profile of UPII in different types of UC and its utility in the diagnostic setting are needed.