Anti-SALL4 Mouse Monoclonal Antibody [clone: 6E3]
Supplier: Diagnostic Biosystems
MOB591-01-RUO MOB591-05-RUO MOB591-RUO
CA76634-228EA 226.56 CAD
CA76634-228 CA76634-230 CA76634-232
Anti-SALL4 Mouse Monoclonal Antibody [clone: 6E3]
Antibodies
Sal-like protein 4 (SALL4) is a zinc-finger transcription factor that serves as a master regulator of embryonic pluripotency and is involved in processes associated with stem cell activities. SALL4 expression in germ cells makes it a useful marker for germ cell tumors such as seminoma, embryonal carcinoma, yolk sac tumors and teratomas. SALL4 expression is also seen in the spermatogonia of normal testis.