Anti-Hepatitis B antigen HBsAg Mouse Monoclonal Antibody [clone: 5C3]
Supplier: Diagnostic Biosystems
MOB570-01-RUO MOB570-05-RUO MOB570-RUO
CA76634-156EA 144.79 CAD
CA76634-156 CA76634-158 CA76634-160
Anti-Hepatitis B antigen HBsAg Mouse Monoclonal Antibody [clone: 5C3]
Antibodies
This antibody stains the cytoplasm of antigen-positive liver cells in patients with Type B viral hepatitis. This antibody reacts with the 'a' determinant present on the HBsAg subtypes ayw1, ayw2, ayw3, ayw4, ayr, adw2, adw4 and adr. It does not react with normal tissues.