Anti-Uroplakin II (UPK2) Rabbit Antibody

Supplier: Diagnostic Biosystems
RP177-01-RUO RP177-05-RUO RP177-RUO
CA76635-204EA 246.83 CAD
CA76635-204 CA76635-206 CA76635-208
Anti-Uroplakin II (UPK2) Rabbit Antibody
Antibodies
Uroplakins (UPs) are a family of transmembrane proteins (UPs Ia, Ib, II and III) that are specific differentiation products of urothelial cells. Uroplakins are markers of terminally differentiated urothelium. Uroplakin II (UPII) is a newly described sensitive marker for urothelial carcinoma (UC). The expression profile of UPII in different types of UC and its utility in the diagnostic setting are needed.
Order Now

SPECIFICATIONS

Antigen symbol UPK2
Antigen name Uroplakin II
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR