Anti-Myelin basic protein Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP052-01-RUO RP052-05-RUO RP052-RUO
CA76635-060EA 200.19 CAD
CA76635-060 CA76635-062 CA76635-064
Anti-Myelin basic protein Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with human myelin basic protein and oligodendrocytes of white matter in brain and spinal cord. Grey matter stains weakly. This antibody stains peripheral nerves also.