Anti-Alpha-1-antitrypsin Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP048-01-RUO RP048-05-RUO RP048-RUO
CA76635-048EA 127.93 CAD
CA76635-048 CA76635-050 CA76635-052
Anti-Alpha-1-antitrypsin Rabbit Polyclonal Antibody
Antibodies
The antibody is specific against human α-1-antitrypsin. This antibody stains macrophages in human tissues and does not stain any other cell type.