Anti-P504S Rabbit Monoclonal Antibody [clone: 13H4]
Supplier: Diagnostic Biosystems
RMAB078-01-RUO RMAB078-05-RUO RMAB078-RUO
CA76634-796EA 302.52 CAD
CA76634-796 CA76634-798 CA76634-800
Anti-P504S Rabbit Monoclonal Antibody [clone: 13H4]
Antibodies
P504S has been recently described as a prostate cancer-specific gene that encodes a protein involved in the β-oxidation of branched chain fatty acids. Expression of p504S protein is found in prostatic adenocarcinoma but not in benign prostatic tissue. It stains premalignant lesions of prostate: high-grade prostatic intraepithelial neoplasia (PIN) and atypical adenomatous hyperplasia. P504S can be used as a positive marker for PIN.