Anti-SOX-10 (melanoma marker) Rabbit Monoclonal Antibody [clone: EP268]
Supplier: Diagnostic Biosystems
RMAB077-01-RUO RMAB077-05-RUO RMAB077-RUO
CA76634-790EA 139.97 CAD
CA76634-790 CA76634-792 CA76634-794
Anti-SOX-10 (melanoma marker) Rabbit Monoclonal Antibody [clone: EP268]
Antibodies
SOX-10, a member of the sex-determining region Y-related HMG-box family, is a transcription factor that is expressed in pre-migratory and migrating neural crest cells. Following migration it remains expressed in cells destined for glial, schwannian, and melanocytic differentiation. SOX10 is widely expressed in normal human tissues including melanocytes and breast tissue. SOX10 is also an important marker in malignant tumors such as melanoma, and breast carcinomas of basal-like and triple negative type, gliomas, and benign tumors such as schwannomas. More importantly, SOX10 has been shown toMore Product Information