Anti-HER2/neu (c-erb-2) Rabbit Monoclonal Antibody [clone: SP3]
Supplier: Diagnostic Biosystems
RMAB008-01-RUO RMAB008-05-RUO RMAB008-RUO
CA76634-720EA 258.87 CAD
CA76634-720 CA76634-722 CA76634-724
Anti-HER2/neu (c-erb-2) Rabbit Monoclonal Antibody [clone: SP3]
Antibodies
This antibody recognizes a c-erbB-2 protein, which is a receptor tyrosine kinase of the c-erbB family. It is closely related in structure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a subset of breast and other adenocarcinomas, as well as transitional cell carcinomas. In the case of breast cancer, expression determined by immunohistochemistry has been shown to be associated with poor prognosis.