Anti-Ki-67 antigen Rabbit Monoclonal Antibody [clone: SP6]
Supplier: Diagnostic Biosystems
RMAB004-01-RUO RMAB004-05-RUO RMAB004-RUO
CA76634-708EA 195.65 CAD
CA76634-708 CA76634-710 CA76634-712
Anti-Ki-67 antigen Rabbit Monoclonal Antibody [clone: SP6]
Antibodies
Ki-67 is a nuclear protein, which is expressed in proliferating cells. Ki-67 is preferentially expressed during late G1-, S-, M-, and G2-phases of the cell cycle, while cells in the G0 (quiescent) phase are negative for this protein.