Anti-Adipophilin Mouse Monoclonal Antibody [clone: DBM15.60]
Supplier: Diagnostic Biosystems
MOB534-01-RUO MOB534-05-RUO MOB534-RUO
CA76634-018EA 183.98 CAD
CA76634-018 CA76634-020 CA76634-022
Anti-Adipophilin Mouse Monoclonal Antibody [clone: DBM15.60]
Antibodies
Recognizes a protein of 48 kDa, which is identified as Adipophilin. It belongs to the perilipin family, members of which coat intracellular lipid storage droplets. This protein is associated with the lipid globule surface membrane material, and maybe involved in development and maintenance of adipose tissue. However, it is not restricted to adipocytes as previously thought, but is found in a wide range of cultured cell lines, including fibroblasts, endothelial and epithelial cells, and tissues, such as lactating mammary gland, adrenal cortex, Sertoli and Leydig cells, and hepatocytes in alcoholic liver cirrhosis, suggesting that it may serve as a marker of lipid accumulation in diverse cell types and diseases.