Anti-GCDFP-15 Mouse Monoclonal Antibody [clone: DBM15.52]
Supplier: Diagnostic Biosystems
MOB526-01-RUO MOB526-05-RUO MOB526-RUO
CA76634-012EA 165.22 CAD
CA76634-012 CA76634-014 CA76634-016
Anti-GCDFP-15 Mouse Monoclonal Antibody [clone: DBM15.52]
Antibodies
It recognizes a protein of 15 kDa, identified as Gross cystic disease fluid protein 15 (GCDFP-15). It is a major protein component of benign breast gross cysts. It is a known marker of breast cancer, as it is found in approximately 50% of all breast cancer specimens. GCDFP-15, also known as PIP, for prolactin inducible protein, is a prolactin and androgen controlled protein. This antibody is useful in the identification of metastatic breast carcinoma, or fluid analysis.