Anti-PD-1 Mouse Monoclonal Antibody [clone: EH33]
Supplier: Diagnostic Biosystems
MOB573-01-RUO MOB573-05-RUO MOB573-RUO
CA76634-168EA 168.65 CAD
CA76634-168 CA76634-170 CA76634-172
Anti-PD-1 Mouse Monoclonal Antibody [clone: EH33]
Antibodies
The programmed death receptor 1 (PD-1) protein is a cell-surface receptor on certain lymphocytes that, with its ligand programmed death ligand 1 (PD-L1), helps to down-regulate immune responses. Many cancer types express PD-L1 and evade immune recognition via the PD-1/PD-L1 interaction. Precision therapies targeting the PD-1/PD-L1 pathway have the potential to improve response and thereby offer a novel treatment avenue to some patients with cancer.