Anti-Podoplanin Mouse Monoclonal Antibody [clone: D2-40]
Supplier: Diagnostic Biosystems
MOB558-01-RUO MOB558-05-RUO MOB558-RUO
CA76634-102EA 292.96 CAD
CA76634-102 CA76634-104 CA76634-106
Anti-Podoplanin Mouse Monoclonal Antibody [clone: D2-40]
Antibodies
Podoplanin is a 38 kDa membrane mucoprotein originally detected on the surface of rat podocytes .The antibody against podoplanin (D2-40) reacts to a 40 kDA sialogylcoprotein found on lymphatic endothelium, fetal testis, and on the surface of testicular germ cell tumors. Podoplanin can be used to assist pathologists in identifying lymphatic endothelium to determine lymphatic invasion and also can be used to help differentiate mesothelioma from adenocarcinoma.