Anti-KU (P70) Mouse Monoclonal Antibody [clone: H-6]
Supplier: Diagnostic Biosystems
MOB557-01-RUO MOB557-05-RUO MOB557-RUO
CA76634-096EA 166.94 CAD
CA76634-096 CA76634-098 CA76634-100
Anti-KU (P70) Mouse Monoclonal Antibody [clone: H-6]
Antibodies
This antibody reacts with a 70 kD protein known as Ku (p70).The Ku protein is localized in the nucleus and is composed of subunits referred to as Ku-70 (or p70) and Ku-86 or (p86). Both subunits of the Ku protein have been cloned, and a number of functions have been proposed for Ku, including cell signaling, DNA replication and transcriptional activation. Ku is involved in Pol II-directed transcription by virtue of its DNA binding activity, serving as the regulatory component of the DNA-associated protein kinase that phosphorylates Pol II and transcription factor Sp.