Anti-Caldesmon Mouse Monoclonal Antibody [clone: hHCD]
Supplier: Diagnostic Biosystems
MOB556-01-RUO MOB556-05-RUO MOB556-RUO
CA76634-090EA 144.79 CAD
CA76634-090 CA76634-092 CA76634-094
Anti-Caldesmon Mouse Monoclonal Antibody [clone: hHCD]
Antibodies
This antibody is specific to high molecular weight (120 to 150 kDa) h-caldesmon. Caldesmon is involved in smooth muscle and non-muscle contraction. Two closely related variants of human caldesmon have been identified. The h-caldesmon variant is predominantly expressed in smooth muscle, whereas l-caldesmon (71 to 80 kDa) is found in non-muscle tissue and cells.