Anti-Thyroid peroxidase Mouse Monoclonal Antibody [clone: MoAb47]
Supplier: Diagnostic Biosystems
MOB418-01-RUO MOB418-05-RUO MOB418-RUO
CA76633-838EA 231.66 CAD
CA76633-838 CA76633-840 CA76633-842
Anti-Thyroid peroxidase Mouse Monoclonal Antibody [clone: MoAb47]
Antibodies
Thyroid peroxidase (TPO) is a transmembrane protein of 107 kDa containing a heme prosthetic group. It is present as a dimer on the apical surface of thyroid follicular cells. TPO is the primary enzyme involved in thyroid hormone synthesis. Malignant thyroid tumors exhibit an anomaly in TPO resulting in lower affinity for anti-TPO. This antibody may aid in the differentiation between benign and malignant thyroid tumors.