Anti-Amyloid beta protein Mouse Monoclonal Antibody [clone: BAM-10]
Supplier: Diagnostic Biosystems
MOB410-01-RUO MOB410-05-RUO MOB410-RUO
CA76633-826EA 144.79 CAD
CA76633-826 CA76633-828 CA76633-830
Anti-Amyloid beta protein Mouse Monoclonal Antibody [clone: BAM-10]
Antibodies
This antibody reacts with amyloid protein. This antibody stains amyloid plaques within the cortex, and amyloid deposits in blood vessels. β-amyloid deposits are also detected in Lewy body dementia, Down syndrome, amyloidosis and in Gram-Parkinson dementia complex. The presence of a large number of neuritic plaques (senile) and neurofibrillary tangles in the cerebral cortex is used as a pathological marker for a disease state and presents the major criterion for the diagnosis of Alzheimer's disease at autopsy.